The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138477.2 | NP_612486.2 | 24 - 25 | 3322 | 123 |
XM_005254176.4 | XP_005254233.1 | 24 - 25 | 3237 | 123 | |
XM_011521270.2 | XP_011519572.1 | 24 - 25 | 3262 | 123 | |
XM_011521271.2 | XP_011519573.1 | 24 - 25 | 3259 | 123 | |
XM_011521274.2 | XP_011519576.1 | 21 - 22 | 2308 | 123 | |
GenBank mRNA | AF525398.1 | - | 24 - 25 | 3319 | 123 |
BC001092.2 | - | 16 - 17 | 1777 | 123 | |
BC008333.1 | - | 16 - 17 | 1777 | 123 | |
BC008334.1 | - | 16 - 17 | 1777 | 123 | |
BC052568.1 | - | 24 - 25 | 3210 | 123 | |
BC066640.1 | - | 24 - 25 | 3210 | 123 |