The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175871.3 | NP_787067.2 | 1 - 2 | 218 | 66 |
GenBank mRNA | AK092438.1 | - | 1 - 2 | 218 | 66 |
BC068071.1 | - | 1 - 2 | 240 | 66 | |
BC119677.2 | - | 1 - 2 | 214 | 66 |