The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176819.3 | NP_789789.2 | 4 - 5 | 1034 | 63 |
XM_005272670.1 | XP_005272727.1 | 3 - 4 | 835 | 63 | |
GenBank mRNA | AK094877.1 | - | 2 - 3 | 259 | 63 |
AK294472.1 | - | 3 - 4 | 603 | 63 | |
DA378381.1 | - | 1 - 2 | 128 | 63 |