The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270391.1 | NP_001257320.1 | 5 - 6 | 699 | 145 |
NM_001270392.1 | NP_001257321.1 | 4 - 5 | 477 | 145 | |
NM_177532.4 | NP_803876.1 | 5 - 6 | 699 | 145 | |
NM_201431.2 | NP_958834.1 | 5 - 6 | 609 | 145 | |
XM_011531687.2 | XP_011529989.1 | 5 - 6 | 601 | 145 | |
XM_011531688.1 | XP_011529990.1 | 6 - 7 | 675 | 145 | |
XM_011531689.2 | XP_011529991.1 | 5 - 6 | 601 | 145 | |
XM_011531690.2 | XP_011529992.1 | 4 - 5 | 3235 | 145 | |
XM_011531691.1 | XP_011529993.1 | 3 - 4 | 327 | 145 | |
XM_017007838.1 | XP_016863327.1 | 5 - 6 | 601 | 145 | |
GenBank mRNA | AK126346.1 | - | 5 - 6 | 601 | 145 |
AY217664.1 | - | 5 - 6 | 513 | 145 | |
AY217665.1 | - | 4 - 5 | 346 | 145 | |
CR749283.1 | - | 5 - 6 | 699 | 145 |