The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177880.1 | NP_001171351.1 | 3 - 4 | 462 | 93 |
NM_001318773.1 | NP_001305702.1 | 4 - 5 | 587 | 93 | |
NM_177963.3 | NP_808878.1 | 3 - 4 | 371 | 93 | |
XM_006718737.3 | XP_006718800.1 | 5 - 6 | 1222 | 93 | |
XM_011545346.2 | XP_011543648.1 | 6 - 7 | 1394 | 93 | |
XM_011545347.1 | XP_011543649.1 | 3 - 4 | 312 | 93 | |
XM_017018547.1 | XP_016874036.1 | 4 - 5 | 475 | 93 | |
XM_017018548.1 | XP_016874037.1 | 4 - 5 | 403 | 93 | |
GenBank mRNA | AK024280.1 | - | 6 - 7 | 1394 | 93 |
AK289727.1 | - | 3 - 4 | 324 | 93 | |
AK315810.1 | - | 3 - 4 | 463 | 93 | |
BC037406.1 | - | 3 - 4 | 296 | 93 | |
DA187677.1 | - | 4 - 5 | 433 | 93 | |
DA239473.1 | - | 3 - 4 | 371 | 93 |