Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
Chromosome Location
Chr.18: 31376777 - 31415791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134453.2 NP_001127925.1 8 - 9 1138 154
NM_177986.4 NP_817123.1 8 - 9 1138 154
GenBank mRNA AY168788.1 - 8 - 9 1032 154
AY177664.1 - 8 - 9 1138 154
AY227350.1 - 8 - 9 1003 154
AY228236.1 - 8 - 9 1032 154
BC132907.1 - 8 - 9 1026 154
BC132909.1 - 8 - 9 1026 154

View Full Product Details