The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178170.2 | NP_835464.1 | 2 - 3 | 256 | 66 |
XM_011524638.2 | XP_011522940.1 | 3 - 4 | 460 | 66 | |
XM_011524640.2 | XP_011522942.1 | 3 - 4 | 460 | 66 | |
XM_017024499.1 | XP_016879988.1 | 2 - 3 | 266 | 66 | |
XM_017024500.1 | XP_016879989.1 | 2 - 3 | 302 | 66 | |
XM_017024501.1 | XP_016879990.1 | 3 - 4 | 460 | 66 | |
GenBank mRNA | AY242354.1 | - | 2 - 3 | 256 | 66 |
AY267371.1 | - | 2 - 3 | 256 | 66 | |
BC112240.1 | - | 2 - 3 | 290 | 66 | |
BC113705.1 | - | 2 - 3 | 290 | 66 |