Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC36
Entrez Gene ID
339834
Gene Name
coiled-coil domain containing 36
Gene Aliases
CT74
Chromosome Location
Chr.3: 49198428 - 49258104 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135197.1 NP_001128669.1 5 - 6 535 121
NM_178173.3 NP_835467.2 7 - 8 833 121
XM_006713124.3 XP_006713187.1 5 - 6 710 121
XM_011533670.2 XP_011531972.1 4 - 5 544 121
XM_011533672.2 XP_011531974.1 5 - 6 903 121
XM_011533673.2 XP_011531975.1 5 - 6 935 121
GenBank mRNA AK058049.1 - 7 - 8 724 121
AK303033.1 - 5 - 6 535 121
BC036202.1 - 7 - 8 833 121

View Full Product Details