Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
118490
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr.10: 73423579 - 73433561 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024593.1 NP_001019764.1 5 - 6 1130 77
GenBank mRNA AK096884.1 - 5 - 6 700 77
AK309870.1 - 6 - 7 1186 77
BC094693.1 - 5 - 6 1109 77
BC109255.1 - 4 - 5 1043 77
BC109256.1 - 4 - 5 1043 77
CR936824.1 - 5 - 6 1112 77

View Full Product Details