The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178824.3 | NP_849146.1 | 2 - 3 | 414 | 93 |
GenBank mRNA | AK097556.1 | - | 6 - 7 | 1312 | 93 |
AK294387.1 | - | 2 - 3 | 302 | 93 | |
BC035512.2 | - | 2 - 3 | 414 | 93 |