Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR49
Entrez Gene ID
151790
Gene Name
WD repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.3: 167478684 - 167653501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178824.3 NP_849146.1 5 - 6 955 97
GenBank mRNA AK090939.1 - 3 - 4 360 97
AK294387.1 - 5 - 6 843 97
BC035512.2 - 5 - 6 955 97

View Full Product Details