The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098831.1 | NP_001092301.1 | 4 - 5 | 476 | 149 |
NM_178832.3 | NP_849154.1 | 4 - 5 | 565 | 149 | |
XM_011539250.2 | XP_011537552.1 | 3 - 4 | 599 | 149 | |
XM_011539251.2 | XP_011537553.1 | 4 - 5 | 549 | 149 | |
GenBank mRNA | AJ312050.1 | - | 3 - 4 | 468 | 149 |
AJ431726.1 | - | 4 - 5 | 476 | 149 | |
AK289406.1 | - | 4 - 5 | 397 | 149 | |
AL834132.1 | - | 4 - 5 | 458 | 149 | |
BC010230.2 | - | 4 - 5 | 427 | 149 | |
BC040015.1 | - | 4 - 5 | 428 | 149 | |
BC093023.1 | - | 4 - 5 | 469 | 149 | |
DQ338435.1 | - | 4 - 5 | 454 | 149 | |
DQ338436.1 | - | 4 - 5 | 454 | 149 |