The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290341.2 | NP_001277270.1 | 7 - 8 | 1100 | 99 |
NM_001290342.2 | NP_001277271.1 | 6 - 7 | 926 | 99 | |
NM_001290343.1 | NP_001277272.1 | 6 - 7 | 837 | 99 | |
NM_178842.4 | NP_849164.2 | 6 - 7 | 833 | 99 | |
XM_011521355.2 | XP_011519657.1 | 7 - 8 | 1143 | 99 | |
XM_011521357.2 | XP_011519659.1 | 7 - 8 | 1037 | 99 | |
XM_011521358.1 | XP_011519660.1 | 7 - 8 | 921 | 99 | |
XM_017022002.1 | XP_016877491.1 | 6 - 7 | 917 | 99 | |
XM_017022003.1 | XP_016877492.1 | 6 - 7 | 842 | 99 | |
XM_017022004.1 | XP_016877493.1 | 5 - 6 | 753 | 99 | |
GenBank mRNA | BC028703.1 | - | 6 - 7 | 914 | 99 |
BC034970.1 | - | 6 - 7 | 835 | 99 |