The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181536.1 | NP_853514.1 | 6 - 7 | 963 | 103 |
XM_017023201.1 | XP_016878690.1 | 6 - 7 | 1386 | 103 | |
XM_017023202.1 | XP_016878691.1 | 6 - 7 | 1386 | 103 | |
XM_017023203.1 | XP_016878692.1 | 6 - 7 | 1386 | 103 | |
XM_017023204.1 | XP_016878693.1 | 6 - 7 | 1386 | 103 | |
GenBank mRNA | AY164485.1 | - | 6 - 7 | 963 | 103 |