The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271856.1 | NP_001258785.1 | 5 - 6 | 459 | 59 |
NM_181711.3 | NP_859062.1 | 6 - 7 | 693 | 59 | |
XM_005268691.3 | XP_005268748.1 | 6 - 7 | 1861 | 59 | |
XM_011537996.1 | XP_011536298.1 | 5 - 6 | 1639 | 59 | |
GenBank mRNA | BC035500.2 | - | 5 - 6 | 430 | 59 |
BC052967.1 | - | 5 - 6 | 377 | 59 | |
BC053666.1 | - | 6 - 7 | 674 | 59 | |
HY035281.1 | - | 5 - 6 | 393 | 59 |