The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286668.1 | NP_001273597.1 | 13 - 14 | 2823 | 115 |
NM_001286669.1 | NP_001273598.1 | 13 - 14 | 2974 | 115 | |
NM_001286670.1 | NP_001273599.1 | 11 - 12 | 2498 | 115 | |
NM_001323890.1 | NP_001310819.1 | 15 - 16 | 3212 | 115 | |
NM_001323892.1 | NP_001310821.1 | 13 - 14 | 2931 | 115 | |
NM_001323893.1 | NP_001310822.1 | 12 - 13 | 2810 | 115 | |
NM_001323899.1 | NP_001310828.1 | 13 - 14 | 2988 | 115 | |
NM_181806.3 | NP_861522.2 | 14 - 15 | 3095 | 115 | |
XM_017007741.1 | XP_016863230.1 | 15 - 16 | 3171 | 115 | |
XM_017007742.1 | XP_016863231.1 | 12 - 13 | 2871 | 115 | |
XM_017007744.1 | XP_016863233.1 | 11 - 12 | 2678 | 115 | |
GenBank mRNA | AF516672.1 | - | 13 - 14 | 2738 | 115 |
AK294332.1 | - | 11 - 12 | 2460 | 115 | |
AK303733.1 | - | 12 - 13 | 2763 | 115 | |
AK304309.1 | - | 14 - 15 | 3086 | 115 | |
AK316296.1 | - | 14 - 15 | 3076 | 115 | |
AY314787.1 | - | 14 - 15 | 3058 | 115 | |
AY422212.1 | - | 14 - 15 | 3010 | 115 | |
BX640635.1 | - | 9 - 10 | 3926 | 115 |