Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML2A
Entrez Gene ID
169611
Gene Name
olfactomedin like 2A
Gene Aliases
PRO34319
Chromosome Location
Chr.9: 124777158 - 124814891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282715.1 NP_001269644.1 2 - 3 452 79
NM_182487.3 NP_872293.2 5 - 6 1033 79
XM_006716989.2 XP_006717052.1 4 - 5 925 79
GenBank mRNA AB119055.1 - 5 - 6 920 79
AK074164.1 - 3 - 4 584 79
AK074709.1 - 4 - 5 79
AY358185.1 - 5 - 6 79
BC054001.1 - 2 - 3 79

View Full Product Details