Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALHM3
Entrez Gene ID
119395
Gene Name
calcium homeostasis modulator 3
Gene Aliases
FAM26A, bA225H22.7
Chromosome Location
Chr.10: 103472804 - 103479240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129742.1 NP_001123214.1 2 - 3 754 89
GenBank mRNA BC039499.1 - 1 - 2 130 89
BC043367.1 - 2 - 3 754 89

View Full Product Details