Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSC5D
Entrez Gene ID
284297
Gene Name
scavenger receptor cysteine rich family member with 5 domains
Gene Aliases
S5D-SRCRB
Chromosome Location
Chr.19: 55488503 - 55519099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144950.1 NP_001138422.1 3 - 4 450 85
NM_001195267.1 NP_001182196.1 3 - 4 450 85
GenBank mRNA AK092577.1 - 2 - 3 776 85
EU699476.1 - 3 - 4 386 85
EU699477.1 - 3 - 4 386 85

View Full Product Details