The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182603.3 | NP_872409.2 | 2 - 3 | 1071 | 86 |
GenBank mRNA | AK095193.1 | - | 2 - 3 | 301 | 86 |