The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015356.4 | NP_056171.3 | 6 - 7 | 573 | 73 |
NM_182706.4 | NP_874365.3 | 6 - 7 | 573 | 73 | |
GenBank mRNA | AF240677.1 | - | 6 - 7 | 566 | 73 |
AF271734.2 | - | 6 - 7 | 544 | 73 | |
AY062238.1 | - | 6 - 7 | 575 | 73 | |
D63481.3 | - | 6 - 7 | 568 | 73 |