The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181608.1 | NP_853639.1 | 1 - 1 | 110 | 94 |
GenBank mRNA | AB180041.1 | - | 1 - 1 | 198 | 94 |