Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
Chromosome Location
Chr.1: 206109849 - 206117048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000707.3 NP_000698.1 2 - 2 1512 118
GenBank mRNA D31833.1 - 2 - 2 1561 118
L37112.1 - 2 - 2 1447 119

View Full Product Details