The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002060.2 | NP_002051.2 | 2 - 2 | 1176 | 103 |
XM_005270750.2 | XP_005270807.1 | 2 - 2 | 1311 | 103 | |
XM_017001043.1 | XP_016856532.1 | 2 - 2 | 1345 | 103 | |
GenBank mRNA | AK291563.1 | - | 2 - 2 | 1175 | 103 |
BC027889.1 | - | 2 - 2 | 1163 | 103 | |
BC072389.1 | - | 2 - 2 | 1166 | 103 | |
M96789.2 | - | 2 - 2 | 1152 | 103 |