The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_125802.1 | 3 - 3 | 1456 | 94 | |
GenBank mRNA | AF008192.1 | - | 3 - 3 | 1456 | 94 |