The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015969.2 | NP_057053.1 | 2 - 2 | 523 | 142 |
GenBank mRNA | AK026553.1 | - | 3 - 3 | 142 | |
BC047445.1 | - | 3 - 3 | 142 | ||
BC054031.2 | - | 2 - 2 | 522 | 142 |