The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 - 4 | 2007 | 127 |
NM_001127400.1 | NP_001120872.1 | 4 - 4 | 2243 | 127 | |
NM_001127401.1 | NP_001120873.1 | 5 - 5 | 2304 | 127 | |
NM_016061.2 | NP_057145.1 | 3 - 3 | 1946 | 127 | |
XM_017004318.1 | XP_016859807.1 | 5 - 5 | 2542 | 127 | |
XM_017004319.1 | XP_016859808.1 | 4 - 4 | 1980 | 127 | |
XM_017004320.1 | XP_016859809.1 | 4 - 4 | 2481 | 127 | |
XM_017004321.1 | XP_016859810.1 | 4 - 4 | 2173 | 127 | |
XM_017004322.1 | XP_016859811.1 | 4 - 4 | 2404 | 127 | |
GenBank mRNA | AF151885.1 | - | 3 - 3 | 1868 | 127 |
AK025730.1 | - | 4 - 4 | 1882 | 127 |