The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021154.4 | NP_066977.1 | 5 - 6 | 702 | 90 |
NM_058179.3 | NP_478059.1 | 5 - 6 | 702 | 90 | |
GenBank mRNA | AF113132.1 | - | 5 - 6 | 599 | 90 |
AK025460.1 | - | 5 - 6 | 636 | 90 | |
AK295222.1 | - | 5 - 6 | 702 | 90 | |
AK295310.1 | - | 6 - 7 | 591 | 90 | |
AY131232.1 | - | 5 - 6 | 611 | 90 | |
BC000971.1 | - | 5 - 6 | 624 | 90 | |
BC004863.2 | - | 5 - 6 | 633 | 90 | |
BC016645.2 | - | 5 - 6 | 612 | 90 | |
BC018129.1 | - | 5 - 6 | 633 | 90 | |
BT006840.1 | - | 5 - 6 | 566 | 90 |