Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16985920-16986181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5378447 c.3307_3308CC>TT p.P1103L Chr.1: 16986154-16986155 on Build GRCh38
7280519 c.3444C>T p.S1148S Chr.1: 16986018-16986018 on Build GRCh38
4389951 c.3431G>A p.R1144H Chr.1: 16986031-16986031 on Build GRCh38
4489063 c.3426C>T p.I1142I Chr.1: 16986036-16986036 on Build GRCh38
4971552 c.3370C>T p.P1124S Chr.1: 16986092-16986092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053148025 Chr.1: 16986138-16986138 on Build GRCh38
rs1025302607 Chr.1: 16986062-16986062 on Build GRCh38
rs979366216 Chr.1: 16985952-16985952 on Build GRCh38
rs758220660 Chr.1: 16986102-16986102 on Build GRCh38
rs753524964 Chr.1: 16986095-16986095 on Build GRCh38
rs768740101 Chr.1: 16986118-16986118 on Build GRCh38
rs780074532 Chr.1: 16986108-16986108 on Build GRCh38
rs925956683 Chr.1: 16985958-16985958 on Build GRCh38
rs773697361 Chr.1: 16986079-16986079 on Build GRCh38
rs367854366 Chr.1: 16986084-16986084 on Build GRCh38
rs770098952 Chr.1: 16986142-16986142 on Build GRCh38
rs537718060 Chr.1: 16986075-16986075 on Build GRCh38
rs560486316 Chr.1: 16986093-16986093 on Build GRCh38
rs542270701 Chr.1: 16986065-16986065 on Build GRCh38
rs973107762 Chr.1: 16986076-16986076 on Build GRCh38
rs763402432 Chr.1: 16986145-16986145 on Build GRCh38
rs937486329 Chr.1: 16986157-16986157 on Build GRCh38
rs747025503 Chr.1: 16986112-16986112 on Build GRCh38
rs780566020 Chr.1: 16985968-16985968 on Build GRCh38
rs778825537 Chr.1: 16985996-16985996 on Build GRCh38
rs964347301 Chr.1: 16985943-16985943 on Build GRCh38
rs556938808 Chr.1: 16986031-16986031 on Build GRCh38
rs749023724 Chr.1: 16985985-16985985 on Build GRCh38
rs535432261 Chr.1: 16985970-16985970 on Build GRCh38
rs981540371 Chr.1: 16986133-16986133 on Build GRCh38
rs750909936 Chr.1: 16985948-16985948 on Build GRCh38
rs987709491 Chr.1: 16986082-16986082 on Build GRCh38
rs925873631 Chr.1: 16986117-16986117 on Build GRCh38
rs760860724 Chr.1: 16986020-16986020 on Build GRCh38
rs766927390 Chr.1: 16986083-16986083 on Build GRCh38
rs762111359 Chr.1: 16986066-16986066 on Build GRCh38
rs777596557 Chr.1: 16985978-16985978 on Build GRCh38
rs552045038 Chr.1: 16986011-16986011 on Build GRCh38
rs778540667 Chr.1: 16986100-16986100 on Build GRCh38
rs185521359 Chr.1: 16986063-16986063 on Build GRCh38
rs776752651 Chr.1: 16986124-16986124 on Build GRCh38
rs189334432 Chr.1: 16986091-16986091 on Build GRCh38
rs943308193 Chr.1: 16985967-16985967 on Build GRCh38
rs76298930 Chr.1: 16985990-16985990 on Build GRCh38
rs1024514052 Chr.1: 16986032-16986032 on Build GRCh38
rs759006564 Chr.1: 16985963-16985963 on Build GRCh38
rs15786 Chr.1: 16986097-16986097 on Build GRCh38
rs557545457 Chr.1: 16986081-16986081 on Build GRCh38
rs775411172 Chr.1: 16986033-16986033 on Build GRCh38
rs769617510 Chr.1: 16986069-16986070 on Build GRCh38
rs755686764 Chr.1: 16986090-16986090 on Build GRCh38
rs748368992 Chr.1: 16986126-16986126 on Build GRCh38
rs766726551 Chr.1: 16986160-16986160 on Build GRCh38
rs41273151 Chr.1: 16986101-16986101 on Build GRCh38
rs970274162 Chr.1: 16986039-16986039 on Build GRCh38

View Full Product Details