Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53431343-53431610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B10
Entrez Gene ID
3028
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Gene Aliases
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1
UniGene
Hs.171280
Gene Chromosome Location
Chr.X: 53431258-53434376 on Build GRCh38
Transcripts
NM_001037811.2 NM_004493.2

SNP Detail

SNP ID Chromosome Location
rs782783783 Chr.X: 53431482-53431482 on Build GRCh38
rs62626305 Chr.X: 53431445-53431445 on Build GRCh38
rs782718839 Chr.X: 53431518-53431518 on Build GRCh38
rs781984645 Chr.X: 53431379-53431379 on Build GRCh38
rs122461163 Chr.X: 53431450-53431450 on Build GRCh38
rs782119770 Chr.X: 53431467-53431467 on Build GRCh38
rs187642167 Chr.X: 53431502-53431502 on Build GRCh38
rs781911950 Chr.X: 53431485-53431485 on Build GRCh38
rs781794810 Chr.X: 53431542-53431542 on Build GRCh38
rs782733278 Chr.X: 53431436-53431436 on Build GRCh38
rs782047786 Chr.X: 53431434-53431434 on Build GRCh38
rs931676673 Chr.X: 53431505-53431505 on Build GRCh38
rs782156100 Chr.X: 53431497-53431497 on Build GRCh38
rs782299531 Chr.X: 53431380-53431380 on Build GRCh38
rs781933715 Chr.X: 53431410-53431410 on Build GRCh38
rs975865504 Chr.X: 53431415-53431415 on Build GRCh38
rs782410328 Chr.X: 53431395-53431395 on Build GRCh38
rs781948930 Chr.X: 53431460-53431460 on Build GRCh38
rs782541533 Chr.X: 53431552-53431552 on Build GRCh38
rs886041974 Chr.X: 53431556-53431556 on Build GRCh38

View Full Product Details