Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1351916-1352165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6522474 c.81G>A p.M27I Chr.16: 1352130-1352130 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755303199 Chr.16: 1352099-1352099 on Build GRCh38
rs1044660992 Chr.16: 1351992-1351992 on Build GRCh38
rs747247122 Chr.16: 1351995-1351995 on Build GRCh38
rs564853174 Chr.16: 1352092-1352092 on Build GRCh38
rs993454027 Chr.16: 1352036-1352036 on Build GRCh38
rs778042839 Chr.16: 1352110-1352110 on Build GRCh38
rs763764801 Chr.16: 1351974-1351974 on Build GRCh38
rs986694444 Chr.16: 1351955-1351955 on Build GRCh38
rs1020962323 Chr.16: 1352066-1352066 on Build GRCh38
rs779505508 Chr.16: 1351985-1351985 on Build GRCh38
rs756910244 Chr.16: 1351977-1351977 on Build GRCh38
rs866920187 Chr.16: 1352104-1352104 on Build GRCh38
rs113470241 Chr.16: 1352021-1352021 on Build GRCh38
rs765220411 Chr.16: 1351938-1351938 on Build GRCh38
rs758954616 Chr.16: 1352107-1352107 on Build GRCh38
rs8052503 Chr.16: 1352142-1352142 on Build GRCh38
rs532236241 Chr.16: 1351984-1351984 on Build GRCh38
rs751036365 Chr.16: 1352086-1352086 on Build GRCh38
rs562191082 Chr.16: 1351993-1351993 on Build GRCh38
rs972447741 Chr.16: 1351975-1351975 on Build GRCh38
rs984755560 Chr.16: 1352140-1352140 on Build GRCh38
rs754225455 Chr.16: 1351982-1351982 on Build GRCh38
rs137853826 Chr.16: 1352123-1352123 on Build GRCh38
rs763639979 Chr.16: 1352082-1352082 on Build GRCh38
rs753208354 Chr.16: 1352103-1352103 on Build GRCh38
rs890685456 Chr.16: 1352044-1352044 on Build GRCh38
rs752212692 Chr.16: 1351944-1351944 on Build GRCh38
rs776361173 Chr.16: 1352007-1352007 on Build GRCh38
rs747111791 Chr.16: 1352111-1352111 on Build GRCh38
rs759365145 Chr.16: 1352069-1352069 on Build GRCh38
rs767184877 Chr.16: 1352094-1352094 on Build GRCh38
rs779788991 Chr.16: 1352146-1352146 on Build GRCh38
rs989859381 Chr.16: 1352115-1352115 on Build GRCh38
rs897789631 Chr.16: 1352028-1352028 on Build GRCh38
rs772239803 Chr.16: 1351991-1351991 on Build GRCh38
rs775363099 Chr.16: 1352075-1352075 on Build GRCh38
rs964111307 Chr.16: 1351960-1351960 on Build GRCh38
rs545117390 Chr.16: 1352073-1352073 on Build GRCh38
rs886051658 Chr.16: 1351971-1351971 on Build GRCh38
rs867217705 Chr.16: 1351973-1351973 on Build GRCh38
rs938573026 Chr.16: 1352148-1352148 on Build GRCh38
rs543660760 Chr.16: 1351978-1351978 on Build GRCh38
rs547624231 Chr.16: 1352116-1352116 on Build GRCh38
rs574801192 Chr.16: 1351976-1351976 on Build GRCh38
rs914207699 Chr.16: 1351972-1351972 on Build GRCh38
rs1009539040 Chr.16: 1352048-1352048 on Build GRCh38
rs955019316 Chr.16: 1351949-1351949 on Build GRCh38
rs745677704 Chr.16: 1352118-1352118 on Build GRCh38
rs762469473 Chr.16: 1352077-1352077 on Build GRCh38
rs1021014743 Chr.16: 1351943-1351943 on Build GRCh38
rs771120488 Chr.16: 1352114-1352114 on Build GRCh38
rs554707396 Chr.16: 1351962-1351962 on Build GRCh38
rs1030320804 Chr.16: 1352037-1352037 on Build GRCh38
rs779597308 Chr.16: 1352100-1352100 on Build GRCh38
rs928528974 Chr.16: 1351988-1351988 on Build GRCh38
rs527348189 Chr.16: 1352097-1352097 on Build GRCh38
rs1017797000 Chr.16: 1351956-1351956 on Build GRCh38
rs989913303 Chr.16: 1351971-1351971 on Build GRCh38
rs977124651 Chr.16: 1351986-1351986 on Build GRCh38
rs754166478 Chr.16: 1352098-1352098 on Build GRCh38
rs748243075 Chr.16: 1351990-1351990 on Build GRCh38
rs575933499 Chr.16: 1352020-1352020 on Build GRCh38
rs928343311 Chr.16: 1352145-1352145 on Build GRCh38
rs967153657 Chr.16: 1352122-1352122 on Build GRCh38
rs771155803 Chr.16: 1352002-1352002 on Build GRCh38
rs942079497 Chr.16: 1352013-1352013 on Build GRCh38

View Full Product Details