Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52319035-52319302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921896 c.574C>A p.L192M Chr.12: 52319175-52319175 on Build GRCh38
3812309 c.497C>T p.T166I Chr.12: 52319252-52319252 on Build GRCh38
4828275 c.492C>G p.I164M Chr.12: 52319257-52319257 on Build GRCh38
266799 c.464A>G p.N155S Chr.12: 52319285-52319285 on Build GRCh38
4801005 c.561C>T p.H187H Chr.12: 52319188-52319188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764816920 Chr.12: 52319102-52319102 on Build GRCh38
rs758707263 Chr.12: 52319136-52319136 on Build GRCh38
rs201851696 Chr.12: 52319165-52319165 on Build GRCh38
rs745406560 Chr.12: 52319170-52319170 on Build GRCh38
rs760912181 Chr.12: 52319113-52319113 on Build GRCh38
rs755055795 Chr.12: 52319124-52319124 on Build GRCh38
rs2187512 Chr.12: 52319114-52319114 on Build GRCh38
rs780885951 Chr.12: 52319233-52319233 on Build GRCh38
rs377272891 Chr.12: 52319256-52319256 on Build GRCh38
rs769643286 Chr.12: 52319206-52319234 on Build GRCh38
rs534475220 Chr.12: 52319097-52319097 on Build GRCh38
rs778964859 Chr.12: 52319247-52319247 on Build GRCh38
rs751246730 Chr.12: 52319139-52319139 on Build GRCh38
rs752054850 Chr.12: 52319280-52319280 on Build GRCh38
rs756856491 Chr.12: 52319141-52319141 on Build GRCh38
rs774246418 Chr.12: 52319190-52319190 on Build GRCh38
rs541989746 Chr.12: 52319243-52319243 on Build GRCh38
rs771321688 Chr.12: 52319194-52319194 on Build GRCh38
rs11834565 Chr.12: 52319269-52319269 on Build GRCh38
rs755592657 Chr.12: 52319238-52319238 on Build GRCh38
rs960602893 Chr.12: 52319215-52319215 on Build GRCh38
rs763480224 Chr.12: 52319227-52319227 on Build GRCh38
rs775593031 Chr.12: 52319263-52319263 on Build GRCh38
rs768576037 Chr.12: 52319188-52319188 on Build GRCh38
rs778049418 Chr.12: 52319138-52319138 on Build GRCh38
rs780797845 Chr.12: 52319143-52319143 on Build GRCh38
rs764623506 Chr.12: 52319275-52319275 on Build GRCh38
rs1009536233 Chr.12: 52319266-52319266 on Build GRCh38
rs759592588 Chr.12: 52319206-52319206 on Build GRCh38
rs140443749 Chr.12: 52319235-52319235 on Build GRCh38
rs776732631 Chr.12: 52319255-52319255 on Build GRCh38
rs372616735 Chr.12: 52319248-52319248 on Build GRCh38
rs879156879 Chr.12: 52319087-52319087 on Build GRCh38
rs138611799 Chr.12: 52319187-52319187 on Build GRCh38
rs769799083 Chr.12: 52319257-52319257 on Build GRCh38
rs776991198 Chr.12: 52319204-52319204 on Build GRCh38
rs961167840 Chr.12: 52319063-52319063 on Build GRCh38
rs1016289056 Chr.12: 52319179-52319179 on Build GRCh38
rs150975843 Chr.12: 52319224-52319224 on Build GRCh38
rs892377936 Chr.12: 52319105-52319105 on Build GRCh38
rs748080107 Chr.12: 52319204-52319209 on Build GRCh38
rs867909772 Chr.12: 52319080-52319080 on Build GRCh38
rs200690744 Chr.12: 52319252-52319252 on Build GRCh38
rs769339595 Chr.12: 52319172-52319172 on Build GRCh38
rs749213363 Chr.12: 52319244-52319244 on Build GRCh38
rs561722742 Chr.12: 52319246-52319246 on Build GRCh38
rs903552585 Chr.12: 52319078-52319078 on Build GRCh38
rs3741715 Chr.12: 52319191-52319191 on Build GRCh38
rs181439241 Chr.12: 52319229-52319229 on Build GRCh38
rs780962178 Chr.12: 52319155-52319157 on Build GRCh38
rs754022019 Chr.12: 52319118-52319118 on Build GRCh38
rs1040745810 Chr.12: 52319267-52319267 on Build GRCh38
rs753234129 Chr.12: 52319134-52319134 on Build GRCh38
rs773581767 Chr.12: 52319112-52319112 on Build GRCh38
rs765732324 Chr.12: 52319131-52319131 on Build GRCh38
rs762219880 Chr.12: 52319283-52319283 on Build GRCh38
rs145476924 Chr.12: 52319276-52319276 on Build GRCh38
rs765304069 Chr.12: 52319218-52319218 on Build GRCh38
rs140783754 Chr.12: 52319264-52319264 on Build GRCh38

View Full Product Details