Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44149849-44150090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCK
Entrez Gene ID
2645
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
UniGene
Hs.1270
Gene Chromosome Location
Chr.7: 44143213-44189439 on Build GRCh38
Transcripts
NM_000162.3 NM_033507.1 NM_033508.1 XM_017011966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3881216 c.571C>T p.R191W Chr.7: 44149977-44149977 on Build GRCh38
3412082 c.561T>C p.A187A Chr.7: 44149984-44149984 on Build GRCh38
3412081 c.564T>C p.A188A Chr.7: 44149984-44149984 on Build GRCh38
3768390 c.484-9T>A p.? Chr.7: 44150073-44150073 on Build GRCh38
3638909 c.580G>A p.G194R Chr.7: 44149971-44149971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs104894006 Chr.7: 44149992-44149992 on Build GRCh38
rs866388269 Chr.7: 44150040-44150040 on Build GRCh38
rs779555700 Chr.7: 44149960-44149960 on Build GRCh38
rs756145968 Chr.7: 44149999-44149999 on Build GRCh38
rs35606092 Chr.7: 44149925-44149926 on Build GRCh38
rs781077527 Chr.7: 44149988-44149988 on Build GRCh38
rs758066627 Chr.7: 44149939-44149939 on Build GRCh38
rs778462693 Chr.7: 44149934-44149934 on Build GRCh38
rs202001955 Chr.7: 44149957-44149957 on Build GRCh38
rs760051415 Chr.7: 44149880-44149880 on Build GRCh38
rs748161171 Chr.7: 44149946-44149946 on Build GRCh38
rs377355289 Chr.7: 44149984-44149984 on Build GRCh38
rs372792788 Chr.7: 44149962-44149962 on Build GRCh38
rs753331941 Chr.7: 44149927-44149927 on Build GRCh38
rs750053191 Chr.7: 44149938-44149938 on Build GRCh38
rs777964292 Chr.7: 44150005-44150005 on Build GRCh38
rs938623069 Chr.7: 44149943-44149943 on Build GRCh38
rs374774643 Chr.7: 44149932-44149932 on Build GRCh38
rs886042610 Chr.7: 44149976-44149976 on Build GRCh38
rs753141536 Chr.7: 44149966-44149966 on Build GRCh38
rs751279776 Chr.7: 44149986-44149986 on Build GRCh38
rs948353492 Chr.7: 44149922-44149922 on Build GRCh38
rs372818361 Chr.7: 44149871-44149871 on Build GRCh38
rs34389297 Chr.7: 44150030-44150030 on Build GRCh38
rs754722633 Chr.7: 44149987-44149987 on Build GRCh38
rs35786405 Chr.7: 44149940-44149940 on Build GRCh38
rs771313675 Chr.7: 44149959-44149959 on Build GRCh38
rs749386678 Chr.7: 44150014-44150014 on Build GRCh38
rs193922306 Chr.7: 44150006-44150006 on Build GRCh38
rs768577208 Chr.7: 44149941-44149941 on Build GRCh38
rs757978639 Chr.7: 44149983-44149983 on Build GRCh38
rs376050856 Chr.7: 44149971-44149971 on Build GRCh38
rs373262279 Chr.7: 44149974-44149974 on Build GRCh38
rs769959696 Chr.7: 44149952-44149952 on Build GRCh38
rs193922307 Chr.7: 44149985-44149985 on Build GRCh38
rs774711466 Chr.7: 44149960-44149960 on Build GRCh38
rs587780344 Chr.7: 44150025-44150025 on Build GRCh38
rs370966942 Chr.7: 44150030-44150030 on Build GRCh38
rs13306394 Chr.7: 44149945-44149945 on Build GRCh38
rs886039380 Chr.7: 44150015-44150015 on Build GRCh38
rs764676295 Chr.7: 44149973-44149973 on Build GRCh38
rs891625796 Chr.7: 44149954-44149954 on Build GRCh38
rs193922303 Chr.7: 44150039-44150039 on Build GRCh38
rs193922305 Chr.7: 44150016-44150016 on Build GRCh38
rs543467370 Chr.7: 44149884-44149884 on Build GRCh38
rs755858412 Chr.7: 44149938-44149939 on Build GRCh38
rs193922304 Chr.7: 44150021-44150021 on Build GRCh38

View Full Product Details