Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132078342-132078603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs377679619 Chr.X: 132078523-132078523 on Build GRCh38
rs910445814 Chr.X: 132078525-132078525 on Build GRCh38
rs370911852 Chr.X: 132078538-132078538 on Build GRCh38
rs766452194 Chr.X: 132078487-132078487 on Build GRCh38
rs528032957 Chr.X: 132078549-132078549 on Build GRCh38
rs868605726 Chr.X: 132078582-132078582 on Build GRCh38
rs759297206 Chr.X: 132078400-132078400 on Build GRCh38
rs770900557 Chr.X: 132078373-132078373 on Build GRCh38
rs762283064 Chr.X: 132078461-132078461 on Build GRCh38
rs745594201 Chr.X: 132078527-132078527 on Build GRCh38
rs762430974 Chr.X: 132078447-132078447 on Build GRCh38
rs769268395 Chr.X: 132078414-132078414 on Build GRCh38
rs143451711 Chr.X: 132078543-132078543 on Build GRCh38
rs867962651 Chr.X: 132078581-132078581 on Build GRCh38
rs200651852 Chr.X: 132078429-132078429 on Build GRCh38
rs776519156 Chr.X: 132078374-132078374 on Build GRCh38
rs199717232 Chr.X: 132078486-132078486 on Build GRCh38
rs767944826 Chr.X: 132078551-132078551 on Build GRCh38
rs755054807 Chr.X: 132078491-132078491 on Build GRCh38
rs377188822 Chr.X: 132078473-132078473 on Build GRCh38
rs746965522 Chr.X: 132078517-132078517 on Build GRCh38
rs763653682 Chr.X: 132078454-132078454 on Build GRCh38
rs181962233 Chr.X: 132078544-132078544 on Build GRCh38
rs200838126 Chr.X: 132078524-132078524 on Build GRCh38
rs192322553 Chr.X: 132078512-132078512 on Build GRCh38
rs5977623 Chr.X: 132078484-132078484 on Build GRCh38
rs754076102 Chr.X: 132078490-132078490 on Build GRCh38
rs61742429 Chr.X: 132078459-132078459 on Build GRCh38
rs199576118 Chr.X: 132078536-132078536 on Build GRCh38

View Full Product Details