Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181236830-181237103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733671 c.889-2A>C p.? Chr.5: 181237044-181237044 on Build GRCh38
7210766 c.923G>C p.R308P Chr.5: 181237008-181237008 on Build GRCh38
4687442 c.908C>T p.T303M Chr.5: 181237023-181237023 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757035539 Chr.5: 181236936-181236936 on Build GRCh38
rs532677868 Chr.5: 181236884-181236884 on Build GRCh38
rs988088554 Chr.5: 181237040-181237040 on Build GRCh38
rs745899160 Chr.5: 181237066-181237066 on Build GRCh38
rs200351563 Chr.5: 181236896-181236897 on Build GRCh38
rs775380109 Chr.5: 181236942-181236943 on Build GRCh38
rs961925115 Chr.5: 181236941-181236941 on Build GRCh38
rs770159352 Chr.5: 181236956-181236956 on Build GRCh38
rs1014625686 Chr.5: 181236940-181236940 on Build GRCh38
rs980884931 Chr.5: 181236881-181236882 on Build GRCh38
rs375378168 Chr.5: 181237049-181237049 on Build GRCh38
rs749907520 Chr.5: 181237012-181237012 on Build GRCh38
rs879223282 Chr.5: 181237046-181237046 on Build GRCh38
rs995101954 Chr.5: 181236974-181236974 on Build GRCh38
rs773626363 Chr.5: 181236957-181236957 on Build GRCh38
rs11445317 Chr.5: 181236945-181236946 on Build GRCh38
rs769155820 Chr.5: 181236946-181236946 on Build GRCh38
rs774758890 Chr.5: 181236940-181236941 on Build GRCh38
rs763160311 Chr.5: 181236970-181236970 on Build GRCh38
rs763974107 Chr.5: 181236997-181236997 on Build GRCh38
rs757851080 Chr.5: 181237013-181237013 on Build GRCh38
rs138233790 Chr.5: 181236907-181236907 on Build GRCh38
rs548459672 Chr.5: 181237007-181237007 on Build GRCh38
rs769235302 Chr.5: 181236939-181236940 on Build GRCh38
rs771185433 Chr.5: 181236973-181236973 on Build GRCh38
rs199990971 Chr.5: 181236996-181236996 on Build GRCh38
rs969258539 Chr.5: 181236947-181236948 on Build GRCh38
rs747352419 Chr.5: 181236918-181236918 on Build GRCh38
rs3179945 Chr.5: 181236928-181236928 on Build GRCh38
rs201701436 Chr.5: 181236911-181236912 on Build GRCh38
rs755178974 Chr.5: 181236941-181236942 on Build GRCh38
rs200119653 Chr.5: 181236942-181236944 on Build GRCh38
rs1052426517 Chr.5: 181236887-181236887 on Build GRCh38
rs781682699 Chr.5: 181237017-181237017 on Build GRCh38
rs914138733 Chr.5: 181236890-181236890 on Build GRCh38
rs369145352 Chr.5: 181237050-181237050 on Build GRCh38
rs868812402 Chr.5: 181237009-181237009 on Build GRCh38
rs568886432 Chr.5: 181236920-181236920 on Build GRCh38
rs796856618 Chr.5: 181237015-181237015 on Build GRCh38
rs935918035 Chr.5: 181236902-181236902 on Build GRCh38
rs900001120 Chr.5: 181236908-181236908 on Build GRCh38
rs554624223 Chr.5: 181236878-181236878 on Build GRCh38
rs775703198 Chr.5: 181237071-181237071 on Build GRCh38
rs555559914 Chr.5: 181236944-181236945 on Build GRCh38
rs1050173709 Chr.5: 181236915-181236915 on Build GRCh38
rs778452699 Chr.5: 181237011-181237011 on Build GRCh38
rs745416693 Chr.5: 181236945-181236945 on Build GRCh38
rs113679128 Chr.5: 181236951-181236952 on Build GRCh38
rs138356698 Chr.5: 181236983-181236983 on Build GRCh38
rs774472032 Chr.5: 181237065-181237065 on Build GRCh38
rs772027951 Chr.5: 181237067-181237067 on Build GRCh38
rs149248823 Chr.5: 181237022-181237022 on Build GRCh38
rs568646577 Chr.5: 181236951-181236951 on Build GRCh38
rs966226069 Chr.5: 181237062-181237062 on Build GRCh38
rs996295054 Chr.5: 181236912-181236912 on Build GRCh38
rs752585622 Chr.5: 181236994-181236994 on Build GRCh38
rs759561448 Chr.5: 181236982-181236982 on Build GRCh38
rs925242928 Chr.5: 181236883-181236883 on Build GRCh38
rs771097360 Chr.5: 181237060-181237060 on Build GRCh38
rs748478755 Chr.5: 181236952-181236952 on Build GRCh38
rs570339393 Chr.5: 181236927-181236930 on Build GRCh38
rs749563972 Chr.5: 181236926-181236927 on Build GRCh38
rs1025191779 Chr.5: 181236943-181236944 on Build GRCh38
rs762618523 Chr.5: 181236976-181236977 on Build GRCh38
rs185952721 Chr.5: 181236942-181236942 on Build GRCh38
rs906647106 Chr.5: 181236921-181236921 on Build GRCh38
rs757685750 Chr.5: 181236909-181236909 on Build GRCh38
rs770353665 Chr.5: 181237023-181237023 on Build GRCh38
rs924297876 Chr.5: 181236891-181236891 on Build GRCh38
rs753724246 Chr.5: 181236998-181236998 on Build GRCh38
rs894323733 Chr.5: 181236911-181236911 on Build GRCh38
rs111534281 Chr.5: 181236886-181236886 on Build GRCh38
rs776115306 Chr.5: 181236942-181236942 on Build GRCh38

View Full Product Details