Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243274416-243274651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3486259 c.311C>T p.S104L Chr.1: 243274547-243274547 on Build GRCh38
6487962 c.397G>T p.E133* Chr.1: 243274633-243274633 on Build GRCh38
5380280 c.352C>T p.L118F Chr.1: 243274588-243274588 on Build GRCh38
4030239 c.331A>G p.N111D Chr.1: 243274567-243274567 on Build GRCh38
3711292 c.313T>A p.L105I Chr.1: 243274549-243274549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771864549 Chr.1: 243274516-243274519 on Build GRCh38
rs924535655 Chr.1: 243274512-243274512 on Build GRCh38
rs980402684 Chr.1: 243274517-243274517 on Build GRCh38
rs767282352 Chr.1: 243274473-243274473 on Build GRCh38
rs778785206 Chr.1: 243274595-243274595 on Build GRCh38
rs913322544 Chr.1: 243274585-243274585 on Build GRCh38
rs745742205 Chr.1: 243274501-243274501 on Build GRCh38
rs977933821 Chr.1: 243274609-243274609 on Build GRCh38
rs759634630 Chr.1: 243274567-243274567 on Build GRCh38
rs779671745 Chr.1: 243274510-243274510 on Build GRCh38
rs901028030 Chr.1: 243274540-243274540 on Build GRCh38
rs143226730 Chr.1: 243274584-243274584 on Build GRCh38
rs776316200 Chr.1: 243274523-243274523 on Build GRCh38
rs759920387 Chr.1: 243274501-243274508 on Build GRCh38
rs773575477 Chr.1: 243274543-243274543 on Build GRCh38
rs927533506 Chr.1: 243274518-243274518 on Build GRCh38
rs149561407 Chr.1: 243274586-243274586 on Build GRCh38
rs1041269666 Chr.1: 243274536-243274536 on Build GRCh38
rs762360060 Chr.1: 243274457-243274457 on Build GRCh38
rs761439373 Chr.1: 243274529-243274529 on Build GRCh38
rs377473337 Chr.1: 243274600-243274600 on Build GRCh38
rs1042922680 Chr.1: 243274490-243274490 on Build GRCh38
rs764887835 Chr.1: 243274481-243274481 on Build GRCh38
rs537215054 Chr.1: 243274507-243274507 on Build GRCh38
rs772141619 Chr.1: 243274505-243274505 on Build GRCh38
rs758051187 Chr.1: 243274601-243274601 on Build GRCh38
rs768097659 Chr.1: 243274519-243274519 on Build GRCh38
rs371148493 Chr.1: 243274533-243274533 on Build GRCh38
rs746538384 Chr.1: 243274513-243274513 on Build GRCh38
rs767563520 Chr.1: 243274572-243274572 on Build GRCh38
rs766899717 Chr.1: 243274557-243274557 on Build GRCh38
rs557656507 Chr.1: 243274539-243274539 on Build GRCh38
rs752112426 Chr.1: 243274562-243274562 on Build GRCh38
rs758361203 Chr.1: 243274608-243274608 on Build GRCh38
rs780115638 Chr.1: 243274611-243274611 on Build GRCh38
rs1049848957 Chr.1: 243274576-243274576 on Build GRCh38

View Full Product Details