The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025204.1 | NP_001020375.1 | 3 - 4 | 285 | 84 |
GenBank mRNA | BC005915.1 | - | 3 - 4 | 231 | 84 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320651.1 | NP_001307580.1 | 3 - 4 | 313 | 84 |
GenBank mRNA | CN280267.1 | - | 3 - 4 | 190 | 84 |