The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039350.2 | NP_001034439.1 | 2 - 3 | 757 | 66 |
NM_001290252.1 | NP_001277181.1 | 2 - 3 | 459 | 66 | |
NM_001290253.1 | NP_001277182.1 | 2 - 3 | 765 | 66 | |
NM_001936.4 | NP_001927.3 | 2 - 3 | 459 | 66 | |
NM_130797.3 | NP_570629.2 | 2 - 3 | 765 | 66 | |
XM_017011808.1 | XP_016867297.1 | 3 - 4 | 711 | 66 | |
XM_017011809.1 | XP_016867298.1 | 3 - 4 | 474 | 66 | |
XM_017011810.1 | XP_016867299.1 | 3 - 4 | 577 | 66 | |
XM_017011811.1 | XP_016867300.1 | 3 - 4 | 501 | 66 | |
GenBank mRNA | AK293582.1 | - | 2 - 3 | 409 | 66 |
AK294483.1 | - | 2 - 3 | 229 | 66 | |
AK308993.1 | - | 2 - 3 | 460 | 66 | |
AK309760.1 | - | 2 - 3 | 561 | 66 | |
BC035912.1 | - | 2 - 3 | 773 | 66 | |
BC150304.1 | - | 2 - 3 | 756 | 66 | |
DA071922.1 | - | 2 - 3 | 409 | 66 | |
M96859.1 | - | 2 - 3 | 506 | 66 | |
M96860.1 | - | 2 - 3 | 353 | 66 |