Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
Chromosome Location
Chr.4: 81426393 - 81471928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300735.1 NP_001287664.1 2 - 3 342 98
NM_152545.2 NP_689758.1 2 - 3 342 98
XM_017007813.1 XP_016863302.1 2 - 3 430 98
GenBank mRNA AK056257.1 - 2 - 3 321 98
AK291459.1 - 2 - 3 290 98
BC036784.1 - 2 - 3 304 98
BC121003.1 - 2 - 3 257 98
BX648337.1 - 2 - 3 255 98

View Full Product Details