Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESYT3
Entrez Gene ID
83850
Gene Name
extended synaptotagmin 3
Gene Aliases
CHR3SYT, E-Syt3, FAM62C
Chromosome Location
Chr.3: 138434573 - 138479942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322831.1 NP_001309760.1 17 - 18 1968 65
NM_001322834.1 NP_001309763.1 17 - 18 1968 65
NM_031913.4 NP_114119.2 17 - 18 1968 65
NR_136409.1 17 - 18 1968 65
XM_017007307.1 XP_016862796.1 16 - 17 1420 65
GenBank mRNA AK289530.1 - 17 - 18 1929 65
BC037292.1 - 16 - 17 2692 65
DQ993202.1 - 17 - 18 1742 65

View Full Product Details