The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256067.1 | NP_001242996.1 | 11 - 12 | 1194 | 119 |
NM_001256068.1 | NP_001242997.1 | 9 - 10 | 1026 | 119 | |
NM_006647.1 | NP_006638.1 | 11 - 12 | 1194 | 119 | |
XM_011518158.2 | XP_011516460.1 | 10 - 11 | 1106 | 119 | |
XM_011518159.1 | XP_011516461.1 | 11 - 12 | 1212 | 119 | |
XM_011518160.1 | XP_011516462.1 | 12 - 13 | 1388 | 119 | |
XM_017014220.1 | XP_016869709.1 | 9 - 10 | 1159 | 119 | |
GenBank mRNA | AB095031.1 | - | 11 - 12 | 1174 | 119 |
AI860821.1 | - | 3 - 4 | 117 | ||
AK098341.1 | - | 9 - 10 | 1119 | 119 | |
AY255769.1 | - | 11 - 12 | 1059 | 119 | |
AY927790.1 | - | 11 - 12 | 1097 | 119 | |
AY927791.1 | - | 9 - 10 | 930 | 119 | |
BC041594.1 | - | 11 - 12 | 1194 | 119 | |
BC110840.1 | - | 11 - 12 | 1178 | 119 |