Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037954.3 NP_001033043.1 16 - 17 1988 82
NM_033425.4 NP_219493.1 12 - 13 1521 82
XM_017018466.1 XP_016873955.1 17 - 18 1948 82
XM_017018467.1 XP_016873956.1 17 - 18 1816 82
XM_017018468.1 XP_016873957.1 12 - 13 1132 82
XM_017018469.1 XP_016873958.1 13 - 14 1157 82
GenBank mRNA AB051522.1 - 12 - 13 1428 82
AK055469.1 - 3 - 4 300 82
AK055899.1 - 10 - 11 1044 82
AK289807.1 - 12 - 13 1497 82
AK294954.1 - 7 - 8 687 82
BC033034.2 - 12 - 13 1480 82
BC041626.1 - 9 - 10 715 82
DQ642016.1 - 16 - 17 1988 82

View Full Product Details