Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLB1
- Entrez Gene ID
-
151056
- Gene Name
- phospholipase B1
- Gene Aliases
-
PLB, PLB/LIP, hPLB
- Chromosome Location
-
Chr.2: 28488579 - 28644135 on Build GRCh38
View Full Product Details