The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201427.1 | NP_001188356.1 | 2 - 3 | 307 | 123 |
NM_015345.3 | NP_056160.2 | 2 - 3 | 371 | 123 | |
XM_006715039.3 | XP_006715102.3 | 2 - 3 | 952 | 123 | |
XM_006715040.3 | XP_006715103.3 | 2 - 3 | 951 | 123 | |
XM_006715042.2 | XP_006715105.1 | 2 - 3 | 2947 | 123 | |
XM_006715043.2 | XP_006715106.1 | 2 - 3 | 324 | 123 | |
XM_006715045.3 | XP_006715108.1 | 2 - 3 | 1539 | 123 | |
XM_006715046.3 | XP_006715109.1 | 2 - 3 | 717 | 123 | |
XM_017010630.1 | XP_016866119.1 | 2 - 3 | 1563 | 123 | |
GenBank mRNA | AB002379.2 | - | 2 - 3 | 307 | 123 |
AK310941.1 | - | 2 - 3 | 314 | 123 | |
AL596701.1 | - | 2 - 3 | 396 | 123 | |
AL833083.2 | - | 2 - 3 | 351 | 123 | |
BC078153.1 | - | 2 - 3 | 596 | 123 |