The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256549.1 | NP_001243478.1 | 8 - 9 | 1026 | 66 |
NM_001257101.1 | NP_001244030.1 | 8 - 9 | 919 | 66 | |
NM_032373.4 | NP_115749.2 | 8 - 9 | 1084 | 66 | |
XM_006718030.1 | XP_006718093.1 | 7 - 8 | 1015 | 66 | |
XM_011540271.2 | XP_011538573.1 | 8 - 9 | 1085 | 66 | |
XM_017016776.1 | XP_016872265.1 | 8 - 9 | 1031 | 66 | |
GenBank mRNA | AK303014.1 | - | 8 - 9 | 1027 | 66 |
AL832003.2 | - | 6 - 7 | 523 | 66 | |
AL832496.1 | - | 8 - 9 | 900 | 66 | |
BC051845.1 | - | 8 - 9 | 894 | 66 |