The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320865.1 | NP_001307794.1 | 6 - 7 | 696 | 92 |
NM_198559.1 | NP_940961.1 | 6 - 7 | 663 | 92 | |
XM_005246539.4 | XP_005246596.1 | 5 - 6 | 660 | 92 | |
XM_005246541.4 | XP_005246598.1 | 4 - 5 | 459 | 92 | |
XM_011511149.2 | XP_011509451.1 | 4 - 5 | 458 | 92 | |
XM_011511150.2 | XP_011509452.1 | 4 - 5 | 424 | 92 | |
XM_017004053.1 | XP_016859542.1 | 4 - 5 | 859 | 92 | |
GenBank mRNA | BC052750.1 | - | 6 - 7 | 663 | 92 |