The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203301.3 | NP_976046.1 | 2 - 3 | 1048 | 150 |
GenBank mRNA | BC053537.1 | - | 2 - 3 | 964 | 150 |
BC068566.1 | - | 2 - 3 | 1020 | 150 | |
BX248772.1 | - | 2 - 3 | 702 | 150 |