The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104554.1 | NP_001098024.1 | 4 - 5 | 851 | 63 |
NM_017705.3 | NP_060175.3 | 4 - 5 | 561 | 63 | |
XM_005254495.4 | XP_005254552.1 | 2 - 3 | 347 | 63 | |
XM_011521720.2 | XP_011520022.1 | 5 - 6 | 722 | 63 | |
XM_011521722.2 | XP_011520024.1 | 5 - 6 | 763 | 63 | |
XM_011521723.2 | XP_011520025.1 | 3 - 4 | 440 | 63 | |
XM_011521727.2 | XP_011520029.1 | 4 - 5 | 616 | 63 | |
XM_017022360.1 | XP_016877849.1 | 7 - 8 | 962 | 63 | |
XM_017022361.1 | XP_016877850.1 | 4 - 5 | 579 | 63 | |
XM_017022362.1 | XP_016877851.1 | 6 - 7 | 882 | 63 | |
XM_017022363.1 | XP_016877852.1 | 3 - 4 | 458 | 63 | |
XM_017022364.1 | XP_016877853.1 | 4 - 5 | 658 | 63 | |
XM_017022365.1 | XP_016877854.1 | 4 - 5 | 616 | 63 | |
XM_017022367.1 | XP_016877856.1 | 4 - 5 | 616 | 63 | |
GenBank mRNA | AK000197.1 | - | 4 - 5 | 851 | 63 |
AY424283.1 | - | 4 - 5 | 851 | 63 | |
BC039234.1 | - | 5 - 6 | 561 | 63 |