Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
Chromosome Location
Chr.1: 17520197 - 17697875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011722.2 NP_001011722.2 9 - 10 947 70
NM_001319837.1 NP_001306766.1 9 - 10 947 70
NM_001319838.1 NP_001306767.1 3 - 4 396 70
NM_001328124.1 NP_001315053.1 3 - 4 396 70
NM_018125.3 NP_060595.3 11 - 12 1180 70
NR_137287.1 11 - 12 1183 70
NR_137288.1 10 - 11 1063 70
XM_005245923.2 XP_005245980.2 10 - 11 1039 70
XM_005245925.2 XP_005245982.2 10 - 11 1027 70
XM_005245927.2 XP_005245984.2 3 - 4 396 70
XM_005245929.2 XP_005245986.2 3 - 4 396 70
XM_006710728.1 XP_006710791.1 10 - 11 1221 70
XM_006710729.1 XP_006710792.1 10 - 11 1036 70
XM_006710731.1 XP_006710794.1 10 - 11 1218 70
XM_011541691.2 XP_011539993.1 9 - 10 1036 70
XM_011541692.2 XP_011539994.1 9 - 10 1045 70
XM_011541693.2 XP_011539995.1 9 - 10 1036 70
XM_011541695.2 XP_011539997.1 9 - 10 1045 70
XM_011541696.2 XP_011539998.1 9 - 10 1045 70
XM_017001617.1 XP_016857106.1 10 - 11 1027 70
XM_017001618.1 XP_016857107.1 4 - 5 314 70
XM_017001619.1 XP_016857108.1 9 - 10 1045 70
XM_017001620.1 XP_016857109.1 9 - 10 1045 70
XM_017001621.1 XP_016857110.1 10 - 11 1163 70
XM_017001622.1 XP_016857111.1 10 - 11 1163 70
GenBank mRNA AB046846.1 - 11 - 12 1180 70
AK092020.1 - 3 - 4 393 70
AK126877.1 - 3 - 4 1301 70
AK300174.1 - 7 - 8 711 70
AK300264.1 - 3 - 4 359 70
AK311076.1 - 10 - 11 990 70
BC117171.1 - 9 - 10 937 70
BC143347.1 - 9 - 10 937 70
DA468685.1 - 3 - 4 396 70
DQ139833.1 - 9 - 10 904 70

View Full Product Details