The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019022.3 | NP_061895.3 | 7 - 8 | 626 | 146 |
XM_005266714.2 | XP_005266771.1 | 6 - 7 | 571 | 146 | |
XM_011526033.2 | XP_011524335.1 | 5 - 6 | 489 | 146 | |
XM_011526034.2 | XP_011524336.1 | 7 - 8 | 618 | 146 | |
XM_011526035.2 | XP_011524337.1 | 7 - 8 | 618 | 146 | |
GenBank mRNA | AB058733.1 | - | 7 - 8 | 588 | 146 |
AK000800.1 | - | 1 - 2 | 66 | 146 | |
AK122715.1 | - | 7 - 8 | 618 | 146 | |
AK295551.1 | - | 6 - 7 | 486 | 146 | |
AK307269.1 | - | 6 - 7 | 499 | 146 | |
AK310389.1 | - | 7 - 8 | 808 | 146 | |
AL832341.1 | - | 7 - 8 | 574 | 146 | |
BC007690.2 | - | 6 - 7 | 417 | 146 | |
BC018981.2 | - | 6 - 7 | 417 | 146 | |
BC093792.1 | - | 7 - 8 | 586 | 146 | |
BC093794.1 | - | 7 - 8 | 587 | 146 | |
BC107422.1 | - | 7 - 8 | 622 | 146 | |
BC143400.1 | - | 6 - 7 | 541 | 146 | |
BX647846.1 | - | 7 - 8 | 635 | 146 |